2,2,4,4,6,8,8-Heptamethylnonane ≥95%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
H2696-100ML
H2696-25ML
700002-198EA
191
USD
700002-198
700002-200
2,2,4,4,6,8,8-Heptamethylnonane ≥95%
2,2,4,4,6,8,8-Heptamethylnonane
2,2,4,4,6,8,8-Heptamethylnonane, also known as Isocetane, is an alkane used as a reference in determining the cetane number of diesel.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₆H₃₄ MW: 226.45 g/mol Density: 0.79 Storage Temperature: Ambient |
MDL Number:
MFCD00008856 CAS Number: 4390-04-9 EINECS: 224-506-8 UN: 3295 ADR: 3,III |
Specification Test Results
Assay | 95% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...