Hexyl isobutyrate ≥98%

H1430-500GM
700004-860EA 84.51 USD
700004-860
Hexyl isobutyrate ≥98%
Hexyl isobutyrate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Density: 0,86
Storage Temperature: Ambient
MDL Number: MFCD00027297
CAS Number: 2349-07-7
EINECS: 219-075-8

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR