2,4-Diaminophenol dihydrochloride ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Hydroxy-1,3-Phenylenediammonium Dichloride

DI103-1KG DI103-25GM DI103-100GM
700007-136EA 233 USD
700007-136 700007-140 700007-138
2,4-Diaminophenol dihydrochloride ≥98%
2,4-Diaminophenol dihydrochloride

2,4-Diaminophenol Dihydrochloride, also known as Amidol, is a dihydrogen chloride salt used as a developing agent in photography; it is a colorless crystalline compound.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₀Cl₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00012979
CAS Number: 137-09-7
EINECS: 205-279-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR