Bis(2-ethylhexyl)adipate

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Dioctyl AdipateBis(2-ethylhexyl) adipate

D1246-200LTB D1246-500MLG
700005-794EA 2441 USD
700005-794 700005-796
Bis(2-ethylhexyl)adipate
Bis(2-ethylhexyl) adipate

Dioctyl Adipate is a diester which is used as a plasticizer.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₂H₄₂O₄
MW: 370.57 g/mol
Density: 0.927
Storage Temperature: Refrigerator
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR