Brij® L4

Warning

Synonyms: Brij 30Laureth-4Brij L4Polyoxyethylene (4) Lauryl Ether

BR100-25GM BR100-2.5KG
700012-468EA 89.65 USD
700012-468 700012-466
Brij® L4
Brij® L4

Brij(R) L4 is a polyoxyethylene fatty ether derived from stearyl alcohols designed to emulsify and produce stable dispersions of cosmetic materials. It appears as a colorless to light yellow liquid that may become hazy or form a precipitate.

Formula: (C₂₀H₄₂O₅)n
Density: 0.95
Storage Temperature: Ambient
MDL Number: MFCD00080891
CAS Number: 9002-92-0
EINECS: 500-002-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR