Benzoin ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxy-2-phenylacetophenone

B2408-5GM B2408-100GM B2408-500GM
700004-042EA 181.16 USD
700004-042 700004-040 700004-044
Benzoin ≥98%
Benzoin

Benzoin, a strong solution of benzoin resin in alcohol, is used most often to treat damaged skin. It is usually applied to the skin prior to adhesive bandages and protects the skin. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR