Dihydrocoumarin ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,2-BenzodihydropyroneChroman-2-One

B2392-500GM B2392-25GM B2392-100GM
700008-326EA 70.31 USD
700008-326 700008-324 700008-328
Dihydrocoumarin ≥99%
Dihydrocoumarin

1,2-Benzodihydropyrone is mainly used as a flavor additive in beverages, gelatins, puddings, candy, and other food items. In cosmetics, it's used as a fragrance ingredient and preservative in products such as perfume, moisturizer, soap and cream.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₈O₂
MW: 148.16 g/mol
Density: 1.169
Storage Temperature: Ambient
MDL Number: MFCD00006881
CAS Number: 119-84-6
EINECS: 204-354-9

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR