Bismuth subsalicylate USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Bismuth(III) subsalicylate

B1703-12KG B1703-25GM B1703-2.5KG B1703-125GM B1703-50KGBL B1703-500GM
89078-632EA 4481.09 USD
89078-632 95032-110 95032-114 95032-112 95032-108 89078-630
Bismuth subsalicylate USP
Bismuth subsalicylate

Bismuth Subsalicylate, USP is an antacid medication used to treat temporary discomforts of the stomach and gastrointestinal tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₇H₅BiO₄
Storage Temperature: Ambient
MDL Number: MFCD00085368
CAS Number: 14882-18-9
EINECS: 238-953-1

Order Now

Specification Test Results

Assay 56% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR