Brij® S2

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Brij S2Brij 72Polyoxyethylene (2) stearyl etherSteareth-2Polyethylene glycol octadecyl ether

B1670-2.5KG B1670-12KG B1670-500GM B1670-25GM
700012-422EA 379 USD
700012-422 700012-428 700012-426 700012-420
Brij® S2
Brij® S2

Brij(R) S2 or Polyoxyethylene (2) Stearyl Ether are designed to emulsify and produce stable dispersions of cosmetic materials. It appears as a colorless to light yellow liquid that may become hazy or form a precipitate

Formula: C₁₈H₃₇(OCH₂CH₂)₂OH
MW: 358.60 g/mol
Density: 0.893 g/mL (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00074177
CAS Number: 9005-00-9
EINECS: 500-017-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR