Brij® IC20

Synonyms: Isoceteth-20POE (20) Isohexadecyl Ether

A1792-20LT
700012-396EA 1402.77 USD
700012-396
Brij® IC20
Brij® IC20

Brij(R) IC20 is non-ionic surfactant of the polyethylene glycol family. It is used as a solubilizer and emulsifying agent in foods, cosmetics, and pharmaceuticals, often as an ointment base and also as a research tool.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₃(OCH₂CH₂)₂₀OH
MW: 1123.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080892
CAS Number: 9004-95-9
EINECS: 500-014-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR