Triethyl acetylcitrate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: O-Acetylcitric acid triethyl esterAcetyltriethylcitrateTriethyl O-Acetylcitrate

A1092-100ML
700006-156EA 97.18 USD
700006-156
Triethyl acetylcitrate ≥99%
Triethyl acetylcitrate
Formula: C₁₄H₂₂O₈
MW: 318.32 g/mol
Density: 1,136
Storage Temperature: Ambient
MDL Number: MFCD00049378
CAS Number: 77-89-4
EINECS: 201-066-5

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR