Tributyl acetylcitrate ≥98%

Synonyms: Tributyl O-AcetylcitrateAcetyltributylcitrateO-Acetylcitric acid tributyl ester

A1091-500GM
700006-154EA 189.09 USD
700006-154
Tributyl acetylcitrate ≥98%
Tributyl acetylcitrate
Formula: C₂₀H₃₄O₈
MW: 402.49 g/mol
Density: 1,05
Storage Temperature: Ambient
MDL Number: MFCD00043554
CAS Number: 77-90-7
EINECS: 201-067-0

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR