Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorescent Calcium Indicators and Related Products

Supplier: Biotium
59001 59003 59002 50031 50030 50033 59005 50032 59004 50035 59007 50043-1 50034 59006 50037 50036 50039 50038 50040 50042 50041 50044 50000 50043 50033-1 50002 50045 50001 50004 50003 50006 50005 50008 50007 50009 59100 50050 50011 50000-1 50010 50013 50012 50015 50014 50016 50020 50022 50021 50024 50023 50026 50025 50027 50029
Calcium is an important intracellular messenger ion responsible for the activation and deactivation of numerous biological events in cells

Calcium indicators differ in their Ca2+ dissociation constants (Kd) or Ca2+ response range, excitation/emission wavelengths, spectral shift, and relative fluorescent quantum yields. Therefore, you should select a Ca2+ indicator that best suits your need in consideration of your biological system, instrument settings and any other fluorescent probes that you may use at the same time. Kd values provide an estimate of the detectable Ca2+ concentration range, usually 0.1 Kd to 10 Kd. However, Kd values in cells could differ significantly from in vitro determined Kd values due to differences in ionic strength, pH, viscosity and Ca2+ buffering by intracellular lipids and proteins.

Biotium offers a number of Ca2+ indicators in both membrane-impermeant salt forms and membrane-permeant AM ester forms. The salt forms of the indicators are water-soluble and can be loaded into cells via microinjection. The AM esters of the indicators are membrane-permeant and thus can be loaded into cells by incubation of the cell or tissue preparation in a buffer containing the AM ester. The AM esters themselves do not bind Ca2+. However, once they have entered the cells, they are hydrolyzed by intracellular esterases into the parent Ca2+ indicators, thus becoming responsive to Ca2+. Biotium also offers related products for calcium studies, including Pluronic F-127, a mild nonionic detergent that can facilitate the loading of AM ester compounds, calcium chelators and calcium ionophores. The calcium calibration kit is used to generate buffers with a range of accurate calcium concentrations for the calibration of fluorescent calcium indicator dyes.

To study the role of calcium in these events, it is essential for one to be able to quantitatively monitor its concentration. The most widely used method of Ca2+ detection is by the use of fluorescent Ca2+ indicators, a technique pioneered by professor Roger Tsien and colleagues (Tsien, R. in Methods in Cell Biology, Vol. 30, Taylor, D.L. and Wang, Y-L, Eds., Academic Press (1989) pp. 127-156).




 

ORDER

Expand All / Collapse All

Calcium Indicators
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Bis-Fura-2, hexapotassium salt 1 mg 50045 89139-234 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, AM ester 10 x 100 μg 50013 89139-180 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, AM ester 1 mg 50014 89139-182 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, AM ester 20 x 50 μg 50016 89139-186 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, AM ester, 1 mM in anhydrous DMSO 1 mL 50015 89139-184 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, pentaammonium salt 1 mg 50010 89139-174 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, pentapotassium salt 1 mg 50011 89139-176 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluo-3, pentasodium salt 1 mg 50012 89139-178 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, AM ester 10 x 100 μg 50033-1 89139-208 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, AM ester 20 x 50 μg 50033 89139-206 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, AM ester 1 mg 50034 89139-210 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, AM ester, 1 mM in anhydrous DMSO 1 mL 50029 89139-198 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, pentaammonium salt - 50025 10018-200 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, pentaammonium salt 1 mg 50030 89139-200 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, pentapotassium salt 1 mg 50031 89139-202 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, pentapotassium salt - 50026 10018-202 Each (500µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fura-2, pentasodium salt 1 mg 50032 89139-204 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Furaptra (Mag-Fura-2), AM ester 20 x 50 μg 50039 89139-220 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Furaptra (Mag-Fura-2), AM ester 10 x 100 μg 50037 89139-216 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Furaptra (Mag-Fura-2), AM ester 1 mg 50038 89139-218 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Furaptra (Mag-Fura-2), tetrapotassium salt 1 mg 50035 89139-212 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Furaptra (Mag-Fura-2), tetrasodium salt 1 mg 50036 89139-214 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Indo-1, AM ester 10 x 100 μg 50043 89139-228 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Indo-1, AM ester 20 x 50 μg 50043-1 89139-230 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Indo-1, AM ester 1 mg 50044 89139-232 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Indo-1, pentaammonium salt 1 mg 50040 89139-222 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Indo-1, pentapotassium salt 1 mg 50041 89139-224 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Indo-1, pentasodium salt 1 mg 50042 89139-226 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhod-2, AM ester 10 x 100 μg 50023 89139-194 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhod-2, AM ester 1 mg 50024 89139-196 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhod-2, triammonium salt 1 mg 50020 89139-188 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhod-2, tripotassium salt 1 mg 50021 89139-190 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhod-2, trisodium salt 1 mg 50022 89139-192 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.


Ion Chelators
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
5,5-Dibromo BAPTA, tetrapotassium salt 100 mg 50004 89139-162 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5,5-Difluoro BAPTA, AM ester 25 mg 50005 89139-164 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5,5-Difluoro BAPTA, tetrapotassium salt 100 mg 50006 89139-166 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5,5-Dimethyl BAPTA, AM ester 25 mg 50007 89139-168 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5,5-Dimethyl BAPTA, tetrapotassium salt 100 mg 50008 89139-170 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5-Methyl-5'-nitro BAPTA, tetrapotassium salt 10 mg 50009 89139-172 Each (10mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

BAPTA, AM ester 25 mg 50000 89139-152 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

BAPTA, AM ester 20 X 1 mg 50000-1 89139-154 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

BAPTA, tetracesium salt 1 g 50001 89139-156 Each (1g) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

BAPTA, tetrapotassium salt 1 g 50002 89139-158 Each (1g) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

BAPTA, tetrasodium salt 1 g 50003 89139-160 Each (1g) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Tpen 100 mg 59003 89139-286 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.


Ionophores
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
4-Bromo A-23187, free acid 1 mg 59006 89139-292 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

A-23187, free acid 1 mg 59001 89139-282 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Ionomycin 1 mg 59007 89139-294 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.


Related Products
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Calcium Calibration Kit 8 assays 59100 89139-296 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DMNP EDTA (caged calcium), tetrapotassium salt 10 mg 50050 10018-204 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

EDC (EDAC) 100 mg 59002 89139-284 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Pluronic F-127, 10% in water 30 mL 59005 89139-290 Each (30ml) Retrieving Restricted
Pluronic F-127 20% in DMSO 1 mL 59004 89139-288 Each (1ml) Retrieving Restricted
Probenecid, sodium salt, water soluble 10 mg 50027 89410-769 Pack of 10 (77mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty