Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorescent Mitochondrial Membrane Potential Dyes

Supplier: Biotium
70017 70016 70005 70018 70012 70015 70014 70011 70055 70054 70010 30001 30019
Biotium offers novel and classic dyes for measuring mitochondrial membrane potential.

  • Novel, classic dyes
  • Measure mitochondrial membrane potential

Loss of mitochondrial membrane potential is a hallmark of apoptosis

MitoView™ Green and Nonyl Acridine Orange (NAO) are green fluorescent, non-potentiometric mitochondrial membrane dyes. Cell staining with non-potentiometric dyes relies on mitochondrial mass, not membrane potential. Thus, these dyes can be used to stain mitochondria in both live cells and fixed cells with green fluorescence, and to visualize mitochondria after depolarization.

Diminished cellular glutathione (GSH) level occurs early in apoptosis due to GSH efflux from mitochondria. The MCB Glutathione Detection Kit is based on monochlorobimane (MCB), which reacts with thiols to form a blue fluorescent product allowing fluorometric quantitation of GSH in cell lysates.

It is an early event preceding phosphatidylserine externalization and coinciding with caspase activation. Biotium offers novel and classic dyes for measuring mitochondrial membrane potential. MitoView™633 is a novel mitochondrial membrane potential dye with far red fluorescence (Ex/Em 622/648 nm), which makes it useful for multi-color fluorescence experiments. In healthy cells, JC-1 dye aggregates in mitochondria as a function of membrane potential, resulting in red fluorescence. Conversely, in apoptotic and necrotic cells with diminished mitochondrial membrane potential, JC-1 exists in a green fluorescent monomeric form in the cytosol, allowing of cell viability to be assessed by measuring the ratio of red to green fluorescence. Rhodamine 123 is a green fluorescent mitochondrial commonly used for flow cytometry measurement of mitochondrial membrane potential. TMRE and TMRM are cell permeable ethyl and methyl esters of tetramethylrhodamine, a red fluorescent dye that accumulates in active mitochondria. DASPEI is a red fluorescent potentiometric mitochondrial dye that has been used in no-wash assays for high content screening. DiIC1(5) is a deep/far red carbocyanine dye which has been used to measure mitochondrial membrane potential in apoptotic cells.

Ordering information: Kit size based on flow cytometry assay or 96-well plate assay.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DASPEI 100 mg 70018 89139-394 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DiIC1(5) 100 mg 70015 89139-388 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

JC-1 (chloride salt) 5 mg 70011 89139-380 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

JC-1 (iodide salt) 5 mg 70014 89139-386 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

JC-1 Mitochondrial Membrane Potential Detection Kit 100 assays 30001 89138-884 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

MCB Glutathione Detection Kit 100 assays 30019 89138-934 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

MitoView™ 633 20 x 50 µg 70055 89139-466 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

MitoView™ Green 20 x 50 µg 70054 89139-464 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Nonyl Acridine Orange (NAO) 50 mg 70012 89139-382 Each (50mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhodamine 123 50 mg 70010 89139-378 Each (50mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Tetramethylrhodamine ethyl ester, perchlorate (TMRE) 25 mg 70016 89139-390 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Tetramethylrhodamine methyl ester, perchlorate (TMRM) 25 mg 70017 89139-392 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

TMRE, 2 mM IN DMSO 0.5 mL 70005 10119-188 Each (500µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty