Chemical-Resistant Printer Labels, Brady
Supplier: Brady
60093 102729 60461
490006-510EA 47.12 USD
490006-510 89125-028 89125-030
Chemical-Resistant Printer Labels, Brady
Labels
Chemical and stain resistant labels for slides and cassettes.
General Labels for Straws, Petri Dishes, Bottles, Flasks, Microplates, Bags, and more:
B-427 Straw label for wrapping, working temperatures from –196 to 80 °C (–321 to 176 °F).