Would you like to log in again?
GlobalMark® Tape, B-515, Brady Worldwide®
Supplier: Brady Worldwide
GlobalMark tapes are easy to install and very durable. Brady's B-515 Static Cling Vinyl is designed for use in cartridge form with the GlobalMark printer. Thickness (PSTC-33): 0.006" (0.15 mm) Abrasion Resistance (Method 5306 of U.S. Federal Test Method Std. No. 191A): CS-10 wheels, 250 g wts. The printed legend withstands up to 800 cycles. Minimum Application Temperature: 50°F (10°C). Service Temperature: -40°F to 158°F (-40°C to 70°C). Average Outdoor Durability: Not recommended for outdoor use.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Size Material For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
4" x 50' Vinyl GlobalMark Printer Clear 76690 89007-678 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty