Would you like to log in again?
Adapter, 75 Degree Angle, Ace Glass Incorporated
Supplier: ACE GLASS, INC.
5092-54
This adapter features a standard taper inner joint at the bottom and side, with a #7 Ace-Thred joint at the top.
  • Bottom and side [ST] inner joints
  • Top Ace-Thred
  • Nylon bushing and FETFE o-ring included
  • Fabricated from borosilicate glass
Adapter comes with a 24/40 standard taper inner joint at the bottom and side. The top joint has a #7 Ace-Thred. Supplied complete with 89051-122 nylon bushing and an 89060-958 FETFE o-ring.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
75 Degree Angle #7 Ace-Thred Adapter 5092-54 89051-316 Each Retrieving
Product available on GSA Advantage® Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty