    Sort by:
  • Search Within Results
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-18, SB-18)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 13177-41-8 MDL Number: MFCD00043218 Molecular Formula: C23H49NO3S Molecular Weight: 419.71 Purity/Analysis Method: 97.0% (N) Form...