H-Series Conductivity Electrodes, Hach
CDW97-K10 CDW97-KP5 CDW97-K1
82026-966EA 698.22 USD
82026-966 82026-962 82026-960
H-Series Conductivity Electrodes, Hach
The conductivity probe is a 4 cell design with 4 platinum band contacts
4 cell conductivity probes give fast and accurate measurement over a wide conductivity range.