Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Precoiled Pipe Markers- Fuel Oil B/Y, NMC

Supplier: NATIONAL MARKER COMP.
A4046 D4046 G4046 F4046 E4046 H4046 C4046 B4046
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold

Can be removed and reapplied in other areas.

Certifications: Meets ANSI A13.1-2007



 

ORDER

Expand All / Collapse All
For Size Height Legend Supplier No. VWR Catalog Number Unit Price Quantity
1.9 - 2.54 cm (3/4 - 1") pipe 1.27 cm (1/2") Fuel Oil B/Y A4046 94003-372 Each Retrieving
11.75 - 14.92 cm (45/8 - 57/8") pipe 3.18 cm (11/4") Fuel Oil B/Y E4046 94012-634 Each Retrieving
15.24 - 20.32 cm (6 - 8") pipe 3.18 cm (11/4") Fuel Oil B/Y F4046 94013-830 Each Retrieving
2.86 - 6.03 cm (11/8 - 23/8") pipe 1.9 cm (3/4") Fuel Oil B/Y B4046 94004-146 Each Retrieving
20.32 - 25.4 cm (8 -10") pipe 6.35 cm (21/2") Fuel Oil B/Y G4046 94014-376 Each Retrieving
25.4 cm (10") pipe 8.89 cm (31/2") Fuel Oil B/Y H4046 94015-156 Each Retrieving
6.35 - 8.26 cm (21/2 - 31/4") pipe 3.18 cm (11/4") Fuel Oil B/Y C4046 94005-562 Each Retrieving
8.57 - 11.43 cm (32/5 - 41/2") pipe 3.18 cm (11/4") Fuel Oil B/Y D4046 94009-482 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty