GC-2010 Plus Gas Chromatograph, Shimadzu
220-94846-42
89217-154EA 21645.56 USD
89217-154
GC-2010 Plus Gas Chromatograph, Shimadzu
Out of the box, the GC-2010 Plus is ready to perform high-speed GC utilizing the latest in ultra narrow-bore capillary columns
The fourth generation Advanced Flow Controller delivers up to 1200 mL/min, 140 psi of pressure and supports split ratios of 9999:1. The GC oven is capable of 20 temperature program ramps and hold steps, and features rapid cooling: 300º C to 50º C <3.5 minutes. In addition, the GC-2010 Plus features best-in-class detectors to meet increasing sensitivity demands for trace-level analysis, long-term stability of peak retention times, and excellent precision, even when using highly volatile solvents. Advanced Flow Technology accessories enable application-specific configurations.
Ordering information: Installation included.