Instatherm® Vacuum Drying Apparatus, Abderhalden, with Flange, Ace Glass Incorporated
Supplier: Ace Glass
9630-05
89075-402EA 974.95 USD
89075-402
Instatherm® Vacuum Drying Apparatus, Abderhalden, with Flange, Ace Glass Incorporated
Instatherm® coated drying chamber with viewing strip which allows variable temperatures to 200°C instantly and enables programmed drying
Temperatures monitored by an adjustable depth thermometer, or a thermowell may be inserted for use with a controlled probe. AC/DC up to 40V, 3 amps. Total capacity of drying chamber is 600 mL. 60 mm I.D. flat flange connection (with groove for a silicone o-ring) between desiccant and drying chamber uses a quick release, stainless steel clamp for easy assembly and disassembly.