KingFisher™ Systems Consumables, Thermo Scientific
Supplier: Thermo Scientific
97002070 97002070 97002080 97002080 97002084 97002084
14230-406PK 420.42 USD
CA14230-406 CA14230-404 14230-406 CA14230-402 14230-404 14230-402
KingFisher™ Systems Consumables, Thermo Scientific
Thermo Scientific Consumables for KingFisher™ Systems are designed specifically for use with these instruments
KingFisher™ instrument utilize disposable plastics made of polypropylene. The plastics - tip combs and microplates - are ideal for magnetic particle processing due to their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads.