You Searched For: 2-tert-Butyl-9,10-anthraquinone
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: TCI America
Description: CAS Number: 84-47-9 MDL Number: MFCD00001232 Molecular Formula: C18H16O2 Molecular Weight: 264.32 Purity/Analysis Method: 98.0% (GC) Form: Cr...