Would you like to log in again?
Multiskan GO Microplate Spectrophotometer, Thermo Scientific
Supplier: THERMO SCIENTIFIC - SAN DIEGO
51119300 51119200
The Multiskan GO is a UV/Vis microplate spectrophotometer offering free wavelength selection, therefore being an ideal tool for virtually any photometric research application, such as DNA, RNA, protein analysis and more
It offers exceptional usability through its intuitive, user-friendly internal software and comprehensive Thermo Scientific SkanIt Software, both available with multiple language options. An automatic power save function reduces energy consumption. Additionally, the Multiskan GO unit meets the RoHS (Restriction of Hazardous Substances) directive. Plate shaking motion is linear.
The Multiskan GO spectrophometer has been designed to deliver first-rate performance and high quality results with minimal user effort. At instrument start up, extensive automatic self diagnostics verifies all major instrument functionalities. Continuous runtime control of optical and mechanical functions also ensures stable day-to-day and year-afteryear performance and reliability.
It measures 96-and 384-well plates, various types of cuvettes and the Thermo Scientific µDrop™ Plate extending the capabilities of the Multiskan GO to include microliter-scale measurements. See catalog number 89201-150.
ORDER
The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.
SPECIFICATIONS
Light Source
Scan speed
Accuracy
Wavelength Selection
Weight (kg)
Bandwidth
Cuvettes
W×D×H
Linearity
Mains Voltage (V/Hz)
Max. Power Consumption (W)
Measurement Time
Power consumption
Precision
Read-out range
USB output
User Interface
Wavelength Range
Plate Format
Incubation Range
ACCESSORIES
Expand All / Collapse All
Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
MIB Installation and Training for Multiskan GO 51119200INST 89168-302 Each Retrieving
MIB Installation and Training for Multiskan GO with Incubator and Cuvette 51119300INST 89168-304 Each Retrieving
Extended Warranty EWMIB10 89184-750 Each Retrieving
Warranty Plus WPMIB10 89184-768 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty