Vertex™ Microdenier Cleanroom Wipers, ITW Texwipe
TX59 TX3052 TX3059
TWTX59CS 397.02 USD
TWTX59 TWTX3052 TWTX3059
Vertex™ Microdenier Cleanroom Wipers, ITW Texwipe
These 100% polyester, sealed-edge, microdenier wipers are processed in a hands-free micro-environment on the Vertex™ manufacturing system
Vertex Sterile wipers are triple-bagged for sterility control. The wipers are sterile validated according to AAMI Guidelines and Certificates of Irradiation and Compliance are attached to each case.
The microdenier yarn has four times the number of filaments and twice the surface area in the yarn bundle compared to standard polyester wipers which increases wiping efficiency. This provides superior wiping and cleaning characteristics including the removal of bacteria from hard surfaces. They do not leave behind a barrier layer and are able to capture more particles within the fiber. The microdenier wipers can be used on scratch sensitive surfaces and are effective at disinfectant application and removal. They are manufactured in an ISO Class 3 environment with a small physical footprint which provides for a more sustainable process. Made in the USA, Vertex wipers are tested quarterly for cytotoxicity and endotoxins.
Ordering information: Vertex wipers are also available in the High Durability Series and the High Sorption Series. The High Sorption Series wipers are available dry, pre-wetted, sterile and nonsterile. This allows for single product qualification.