BioCision® CoolProbe™ M, traceable temperature monitor
Supplier: BioCision
BCS-201
89214-826EA 340.55 USD
89214-826
BioCision® CoolProbe™ M, traceable temperature monitor
Thermometers Digital Thermometers
CoolProbe traceable thermometer with microfuge tube probe fitting provides accurate sample temperature monitoring while cooling in CoolRack M-series thermo-conductive microfuge tube modules
Sampling rate Fast (10 seconds) or Normal (60 seconds), Microcentrifuge tube probe type, LCD digital thermometer, Internal (ambient) or external (probe) temperature display with minimum/maximum memory display and High/low alarm.
For CoolProbe U, with universal probe see 89214-824. For CoolProbe CF with cryovial probe, see 89214-828.
Microfuge tube-shaped probe snugly fits in wells of the CoolRack M modules and displays high, low and current sample temperature, with memory and alarm features. NIST traceable and comes with certification.