Anti-MAGEB4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP6174A
80088-992EA 751.67 USD
80088-992
Anti-MAGEB4 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MAGEB4 (N-term) Reacts with Human

Type: Primary
Antigen: MAGEB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (165–180 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR