Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemical
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: Pan

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Source Supplier No. VWR Catalog Number Unit Price Quantity
1 mg Aequorea victoria 600-102-215 RL600-102-215 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol GFP
Antigen Name Green Fluorescence Protein
Reactivity All Species
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Antibody Type Tag
Host Goat
Isotype IgG
Western Blot Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Antigen Synonyms GFPuv, Green Fluorescent Protein, eGFP
Epitope Full length
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen Recombinant Green Fluorescent Protein (GFP) fusion Protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
Purification purified by immunoaffinity chromatography
Form Lyophilized
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage Temperature -20°C
Shipping Temperature Ambient
Tested Applications Fluoresence ELISA
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty