Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
604-1203 604-1203
RL604-1203EA 220.12 USD
CA604-1203 RL604-1203
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Secondary Goat Anti-IgG F(c) Reacts with Dog (Canine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR