Anti-SARS-CoV Membrane Protein Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A55 100-401-A55
RL100-401-A55EA 464.71 USD
CA100-401-A55 RL100-401-A55
Anti-SARS-CoV Membrane Protein Rabbit Polyclonal Antibody
This antibody has been tested for use in immunofluorescence microscopy, immunoprecipitation, immunoelectron microscopy and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 25 kDa.

Recommended Dilutions: Immunofluorescence: 1:1000; Western Blot: 1:1000; Immunoprecipitation: 1:60; Immunoelectron Microscopy 1:200

Type: Primary
Antigen: SARS-CoV Membrane Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: SARS
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR