Anti-Rat Serum Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
112-4101 112-4101
RL112-4101EA 318.82 USD
RL112-4101 CA112-4101
Anti-Rat Serum Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-RAT SERUM Reacts with Rat

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Rat Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR