Anti-PKM Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-1178 100-1178
RL100-1178EA 1289.92 USD
RL100-1178 CA100-1178
Anti-PKM Goat Polyclonal Antibody
Antibodies
Anti-PYRUVATE KINASE Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation, conjugation and most immunological methods requiring high titer and specificity. Specific conditions should be optimized by user.

Recommended Dilutions: ELISA: 1:50000 - 1:200,000; Western Blot: 1:5000 - 1:20000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PKM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR