Anti-HRP Rat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-5138 100-5138
RL100-5138EA 433.62 USD
RL100-5138 CA100-5138
Anti-HRP Rat Polyclonal Antibody
Antibodies
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:2000 - 1:6000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR