Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P53 Mouse Polyclonal Antibody [clone: BP53-12]

Supplier: Rockland Immunochemical
200-301-174
This antibody is suitable for ChIP, flow cytometry, immunochemistry, immunoblotting and immunoprecipitation.  p53 is the most commonly mutated gene in spontaneously occurring human cancers. Mutations arise with an average frequency of 70%.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; ChIP: 1 µg/µl at 4° o/n; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 1:100;

Type: Primary
Antigen: P53
Clonality: Polyclonal
Clone: BP53-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 200-301-174 RL200-301-174 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol P53
Antigen Name Tumor protein p53
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Clone BP53-12
Host Mouse
Isotype IgG2a kappa
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
ChIP Yes
Antigen Synonyms antigen NY-CO-13, P53, or transformation-related protein 53 (TRP53), LFS1, tumor suppressor p53, BCC7, phosphoprotein p53, TRP53, tumor protein p53
Gene ID 7157
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen This Protein A purified monoclonal antibody was produced by repeated immunizations with recombinant human p53 Protein.
Purification Protein A purified
Form Liquid (sterile filtered)
Storage Buffer 0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Dry Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty