Anti-EXOSC1 Mouse Polyclonal Antibody [clone: BM3]

Supplier: Rockland Immunochemicals
200-301-277 200-301-277
RL200-301-277EA 528.35 USD
RL200-301-277 CA200-301-277
Anti-EXOSC1 Mouse Polyclonal Antibody [clone: BM3]
Antibodies
This antibody is suitable for flow cytometry and frozen section.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: EXOSC1
Clonality: Polyclonal
Clone: BM3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR