Anti-NOTCH1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-405 100-401-405
RL100-401-405EA 518.35 USD
RL100-401-405 CA100-401-405
Anti-NOTCH1 Rabbit Polyclonal Antibody
Antibodies
Anti-Notch-1 Antibody shows reactivity at a 1:1000 dilution by western blot.  A strong response was detected by ELISA against the immunizing peptide. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: NOTCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR