Anti-RELA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-266 100-401-266
RL100-401-266EA 518.16 USD
RL100-401-266 CA100-401-266
Anti-RELA Rabbit Polyclonal Antibody
Antibodies
Anti-phospho NFkB antibody reacts human pS529 p65 and shows minimal reactivity by western blot with non-phosphorylated p65 and minimal reactivity by ELISA against the non-phosphorylated form of the immunizing peptide.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000;

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR