Anti-NFKB Polyclonal Antibody

Supplier: Rockland Immunochemicals
K711 K-711
RLK-711EA 1057.64 USD
CAK-711 RLK-711
Anti-NFKB Polyclonal Antibody
Antibodies
Primary Anti-NFKB Reacts with Pan

Type: Primary
Antigen: NFKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR