Anti-Mouse Serum Goat Monoclonal Antibody [clone: 2D11.H5]

Supplier: Rockland Immunochemicals
110-1101 110-1101
RL110-1101EA 318.83 USD
CA110-1101 RL110-1101
Anti-Mouse Serum Goat Monoclonal Antibody [clone: 2D11.H5]
Antibodies
Primary Goat Anti-MOUSE SERUM Reacts with Mouse

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Mouse Serum
Clonality: Monoclonal
Clone: 2D11.H5
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR