Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MED23 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
Anti-DRIP-130 detects a 130 kDa band by immunoblot analysis using a dilution of 1:1,000. For immunoprecipitation a dilution of 1:100 or 1:200 is suggested; pre-clearing with a non-specific rabbit IgG is helpful to reduce background.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000; Immunoprecipitation: 1:100 - 1:200;
Restore with deionized water (or equivalent); 500 µl

Type: Primary
Antigen: MED23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DRIP130 (Mouse, 897–196), Antiserum 500 µL 100-401-239 RL100-401-239 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol MED23
Antigen Name Mediator Complex Subunit 23
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
ELISA Yes
ImmunoPrecipitation Yes
Antigen Synonyms MRT18, DRIP130, SUR-2, CRSP3, CRSP130, SUR2, ARC130, CRSP133
Gene ID 9439
Concentration 85 mg/ml by Refractometry
Format Antiserum
Cross Adsorption No
Immunogen This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide corresponding to mouse DRIP-130 residues  897-916 (based on human DRIP-130 residue numbering)  crosslinked to KLH.
Purification delipidation and defibrination
Form Lyophilized
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Ambient
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty