Anti-MDH1 Sheep Monoclonal Antibody [clone: WA-1]

Supplier: Rockland Immunochemicals
100-601-145 100-601-145
RL100-601-145EA 1289.92 USD
CA100-601-145 RL100-601-145
Anti-MDH1 Sheep Monoclonal Antibody [clone: WA-1]
Antibodies
This product has been assayed against 1.0 µg of Malate Dehydrogenase (Pig Heart) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: MDH1
Clonality: Monoclonal
Clone: WA-1
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR