Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LYZ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
Anti-LYSOZYME antibody has been assayed against 1.0 µg of Lysozyme (Hen Egg White) in a standard ELISA. A working dilution of 1:1000 to 1:2000 of the reconstitution concentration is suggested for this product.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: LYZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Lysozyme (Chicken Egg White), Antiserum 2 mL 100-4172 RL100-4172 Each (2ml) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

View MSDS SDS
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol LYZ
Antigen Name Lysozyme
Reactivity Chicken
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 17D10
Host Rabbit
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Antigen Synonyms LZM
Gene ID 395708
Concentration 80 mg/ml by Refractometry
Format Antiserum
Cross Adsorption No
Immunogen Lysozyme (Hen Egg White)
Purification delipidation and defibrination
Form Lyophilized
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Ambient
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty