Anti-LPO Sheep Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-601-146 100-601-146
RL100-601-146EA 1289.92 USD
CA100-601-146 RL100-601-146
Anti-LPO Sheep Polyclonal Antibody
Antibodies
Primary Sheep Anti-LACTOPEROXIDASE (Bovine Milk) Reacts with Pan

Recommended Dilutions: ELISA: 1:18000 - 1:75000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: LPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Bovine, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR