Anti-HMGN1 Mouse Polyclonal Antibody [clone: IPO-38]

Supplier: Rockland Immunochemicals
200-301-278 200-301-278
RL200-301-278EA 528.35 USD
RL200-301-278 CA200-301-278
Anti-HMGN1 Mouse Polyclonal Antibody [clone: IPO-38]
Antibodies
Anti-IPO-38 is suitable for ELISA,  immunoprecipitation, western blotting and immunochemistry (frozen and formalin/paraffin). The antibody is reported to recognize a nuclear antigen that is present in the cytoplasm and nuclei of proliferating cells.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 10 µg/ml;

Type: Primary
Antigen: HMGN1
Clonality: Polyclonal
Clone: IPO-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR