Anti-NFKBIB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4186 100-4186
RL100-4186EA 518.16 USD
RL100-4186 CA100-4186
Anti-NFKBIB Rabbit Polyclonal Antibody
Antibodies
This product was assayed by immunoblot and found to be reactive against IkBb at a dilution of 1:1000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: NFKBIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR