Anti-TNFA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-306 109-401-306
RL109-401-306EA 1289.92 USD
RL109-401-306 CA109-401-306
Anti-TNFA Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Human TNF alpha Reacts with Human

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR