Anti-SUMO1 Rabbit Polyclonal Antibody [clone: G3.1]

Supplier: Rockland Immunochemicals
200-401-441 200-401-441
RL200-401-441EA 569.75 USD
CA200-401-441 RL200-401-441
Anti-SUMO1 Rabbit Polyclonal Antibody [clone: G3.1]
Antibodies
This purified polyclonal antibody reacts with human SUMO by western blot and ELISA.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: SUMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR