Anti-DDB2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A10 100-401-A10
RL100-401-A10EA 518.16 USD
CA100-401-A10 RL100-401-A10
Anti-DDB2 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human DDB2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using 293T, and others).  Coimmunoprecipitation of related Proteins has not been tested.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: DDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR