Anti-EP300 Mouse Polyclonal Antibody [clone: AC240]

Supplier: Rockland Immunochemicals
100-301-176 100-301-176
RL100-301-176EA 528.35 USD
CA100-301-176 RL100-301-176
Anti-EP300 Mouse Polyclonal Antibody [clone: AC240]
Antibodies
This product was assayed by immunoblot and found to be reactive against p300 at a dilution of 1:10 to 1:50 followed by reaction with Peroxidase conjugated Affinity Purified anti-Mouse IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: EP300
Clonality: Polyclonal
Clone: AC240
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR